Assay Details
| Gene Symbol: C5H11orf74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286248.1 | NP_001273177.1 | 4 - 5 | 429 | 75 | |
| NR_104453.1 | 3 - 4 | 262 | 75 | |||
| XM_015286996.2 | XP_015142482.2 | 4 - 5 | 721 | 75 | ||
| GenBank mRNA | BU345906.1 | - | 4 - 5 | 429 | 75 | |
| BU444136.1 | - | 3 - 4 | 251 | 75 | ||
| BX265911.3 | - | 2 - 3 | 271 | 75 | ||
| BX265912.4 | - | 3 - 4 | 261 | 75 | ||
| BX930430.1 | - | 4 - 5 | 425 | 75 | ||
| BX930817.1 | - | 4 - 5 | 412 | 75 | ||
| CN228581.1 | - | 3 - 4 | 314 | 75 | ||
| CO635604.1 | - | 5 - 6 | 420 | 75 | ||
More Information
Set Membership: |
|

C5H11orf74
Genomic Map