Assay Details
| Gene Symbol: C5H11orf74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286248.1 | NP_001273177.1 | 6 - 7 | 614 | 117 | |
| NR_104453.1 | 5 - 6 | 447 | 117 | |||
| XM_015286996.2 | XP_015142482.2 | 6 - 7 | 903 | 117 | ||
| GenBank mRNA | BU345906.1 | - | 7 - 8 | 614 | 117 | |
| BU444136.1 | - | 5 - 6 | 436 | 117 | ||
| BX265911.3 | - | 4 - 5 | 456 | 117 | ||
| BX265912.4 | - | 5 - 6 | 445 | 117 | ||
| BX930430.1 | - | 7 - 8 | 610 | 117 | ||
| BX930817.1 | - | 7 - 8 | 597 | 117 | ||
| CO635604.1 | - | 8 - 9 | 605 | 117 | ||
More Information
Set Membership: |
|

C5H11orf74
Genomic Map