Assay Details
| Gene Symbol: DPCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277365.1 | NP_001264294.1 | 5 - 6 | 558 | 61 | |
| NM_001277366.1 | NP_001264295.1 | 5 - 6 | 540 | 61 | ||
| GenBank mRNA | BU353811.1 | - | 5 - 6 | 520 | 61 | |
| BU399154.1 | - | 5 - 6 | 527 | 61 | ||
| BX931687.2 | - | 4 - 5 | 777 | 61 | ||
| BX931759.1 | - | 5 - 6 | 535 | 61 | ||
| BX932380.2 | - | 1 - 2 | 625 | 61 | ||
| BX932644.1 | - | 5 - 6 | 533 | 61 | ||
| BX932782.1 | - | 5 - 6 | 550 | 61 | ||
| BX933238.1 | - | 5 - 6 | 534 | 61 | ||
| CR386779.1 | - | 5 - 6 | 516 | 61 | ||
More Information
Set Membership: |
|

DPCD
Genomic Map