Assay Details
Gene Symbol: RAB19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278002.1 | NP_001264931.1 | 3 - 4 | 481 | 69 | |
NM_001278003.1 | NP_001264932.1 | 3 - 4 | 459 | 69 | ||
GenBank mRNA | BX930148.1 | - | 3 - 4 | 437 | 69 | |
BX931864.1 | - | 3 - 4 | 435 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
