Assay Details
| Gene Symbol: FCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004945743.3 | XP_004945800.1 | 7 - 8 | 4448 | 84 | |
| XM_004945746.3 | XP_004945803.1 | 7 - 8 | 4389 | 84 | ||
| XM_004945747.3 | XP_004945804.1 | 6 - 7 | 4352 | 84 | ||
| XM_015279602.2 | XP_015135088.1 | 8 - 9 | 3238 | 84 | ||
| XM_015279603.2 | XP_015135089.1 | 7 - 8 | 4532 | 84 | ||
| XM_015279604.2 | XP_015135090.1 | 7 - 8 | 4482 | 84 | ||
| XM_015279605.2 | XP_015135091.1 | 6 - 7 | 2366 | 84 | ||
| XM_025141453.1 | XP_024997221.1 | 5 - 6 | 2180 | 84 | ||
| GenBank mRNA | BX931873.1 | - | 1 - 2 | 250 | 84 | |
| CR406783.1 | - | 6 - 7 | 439 | 84 | ||
More Information
Set Membership: |
|

FCN2
Genomic Map