Assay Details
| Gene Symbol: FZD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271940.3 | NP_001258869.1 | 3 - 4 | 753 | 67 | |
| NM_001271941.3 | NP_001258870.1 | 3 - 4 | 593 | 67 | ||
| GenBank mRNA | BX932094.1 | - | 3 - 4 | 721 | 67 | |
| BX932219.1 | - | 3 - 4 | 593 | 67 | ||
More Information
Set Membership: |
|

FZD3
Genomic Map