Assay Details
| Gene Symbol: USP46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004936029.3 | XP_004936086.1 | 8 - 9 | 1350 | 95 | |
| XM_420711.6 | XP_420711.1 | 7 - 8 | 1255 | 95 | ||
| GenBank mRNA | BX932326.1 | - | 5 - 6 | 776 | 95 | |
| BX934081.1 | - | 7 - 8 | 1237 | 95 | ||
More Information
Set Membership: |
|

USP46
Genomic Map