Assay Details
| Gene Symbol: URAH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307950.1 | NP_001294879.1 | 1 - 2 | 97 | 59 | |
| XM_025154196.1 | XP_025009964.1 | 2 - 3 | 729 | 59 | ||
| GenBank mRNA | BX932661.1 | - | 1 - 2 | 87 | 59 | |
| BX934929.1 | - | 1 - 2 | 120 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001307950.1, XM_025154196.1 transcripts.
Set Membership: |
|

URAH
Genomic Map