Assay Details
Gene Symbol: ZHX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015283160.2 | XP_015138646.1 | 2 - 3 | 213 | 75 | |
XM_015283161.2 | XP_015138647.1 | 2 - 3 | 204 | 75 | ||
GenBank mRNA | BX930567.1 | - | 2 - 3 | 152 | 75 | |
BX933542.1 | - | 2 - 3 | 153 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015283160.2, XM_015283161.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
