Assay Details
Gene Symbol: MRPL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_419623.5 | XP_419623.1 | 2 - 3 | 366 | 72 | |
GenBank mRNA | BX930290.1 | - | 2 - 3 | 338 | 72 | |
BX930840.1 | - | 2 - 3 | 347 | 72 | ||
BX934122.1 | - | 2 - 3 | 372 | 72 | ||
BX935429.2 | - | 2 - 3 | 351 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
