Assay Details
| Gene Symbol: NDUFB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277896.1 | NP_001264825.1 | 1 - 2 | 124 | 120 | |
| XM_015285557.2 | XP_015141043.1 | 1 - 2 | 87 | 120 | ||
| XM_025148782.1 | XP_025004550.1 | 1 - 2 | 97 | 120 | ||
| GenBank mRNA | BU289042.1 | - | 1 - 2 | 120 | ||
| BU441306.1 | - | 1 - 2 | 124 | 120 | ||
| BX930061.2 | - | 1 - 2 | 100 | 120 | ||
| BX930214.1 | - | 1 - 2 | 120 | |||
| BX930452.2 | - | 1 - 2 | 72 | 120 | ||
| BX934184.1 | - | 1 - 2 | 70 | 120 | ||
| CN227462.1 | - | 1 - 2 | 124 | 120 | ||
| CR353359.1 | - | 1 - 2 | 118 | 120 | ||
More Information
Set Membership: |
|

NDUFB2
Genomic Map