Assay Details
| Gene Symbol: COA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277642.1 | NP_001264571.1 | 3 - 4 | 553 | 92 | |
| NM_001277643.1 | NP_001264572.1 | 3 - 4 | 392 | 92 | ||
| GenBank mRNA | BU427269.1 | - | 3 - 4 | 359 | 92 | |
| BU436789.1 | - | 2 - 3 | 229 | 92 | ||
| BU466385.1 | - | 3 - 4 | 410 | 92 | ||
| BX929699.1 | - | 1 - 2 | 318 | 92 | ||
| BX929802.2 | - | 1 - 2 | 303 | 92 | ||
| BX929935.2 | - | 3 - 4 | 553 | 92 | ||
| BX930119.1 | - | 2 - 3 | 257 | 92 | ||
| BX931531.1 | - | 1 - 2 | 323 | 92 | ||
| BX931563.2 | - | 3 - 4 | 438 | 92 | ||
| BX934104.1 | - | 2 - 3 | 337 | 92 | ||
| BX935003.1 | - | 2 - 3 | 250 | 92 | ||
More Information
Set Membership: |
|

COA7
Genomic Map