Assay Details
| Gene Symbol: CPSF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277551.1 | NP_001264480.1 | 1 - 2 | 142 | 78 | |
| GenBank mRNA | BX935063.1 | - | 1 - 2 | 151 | 78 | |
More Information
Set Membership: |
|

CPSF4
Genomic Map