Assay Details
Gene Symbol: GSTZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277462.1 | NP_001264391.1 | 7 - 8 | 538 | 57 | |
NM_001277469.1 | NP_001264398.1 | 7 - 8 | 489 | 57 | ||
NM_001277470.1 | NP_001264399.1 | 7 - 8 | 441 | 57 | ||
NR_102408.1 | 6 - 7 | 462 | 57 | |||
XM_015287396.2 | XP_015142882.1 | 8 - 9 | 660 | 57 | ||
XM_015287397.2 | XP_015142883.1 | 8 - 9 | 659 | 57 | ||
GenBank mRNA | BU220196.1 | - | 6 - 7 | 472 | 57 | |
BU240466.1 | - | 7 - 8 | 479 | 57 | ||
BU307172.1 | - | 7 - 8 | 485 | 57 | ||
BU461718.1 | - | 7 - 8 | 501 | 57 | ||
BX930489.1 | - | 7 - 8 | 447 | 57 | ||
BX935175.1 | - | 7 - 8 | 506 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
