Assay Details
| Gene Symbol: ATP5G1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX935386.1 | - | 1 - 2 | 342 | 97 | |
| BX935871.1 | - | 2 - 3 | 128 | 97 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATP5G1| Gene Symbol: ATP5G1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX935386.1 | - | 1 - 2 | 342 | 97 | |
| BX935871.1 | - | 2 - 3 | 128 | 97 | ||
Set Membership: |
|