Assay Details
| Gene Symbol: ATP5G1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001233602.5 | XP_001233603.2 | 4 - 5 | 376 | 95 | |
| XM_418114.6 | XP_418114.3 | 4 - 5 | 393 | 95 | ||
| GenBank mRNA | BX935386.1 | - | 3 - 4 | 594 | 95 | |
| BX935871.1 | - | 4 - 5 | 380 | 95 | ||
More Information
Set Membership: |
|

ATP5G1
Genomic Map