Assay Details
| Gene Symbol: GATC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004934392.3 | XP_004934449.1 | 1 - 2 | 149 | 129 | |
| XM_415269.6 | XP_415269.3 | 1 - 2 | 149 | 129 | ||
| GenBank mRNA | BX930114.1 | - | 1 - 2 | 120 | 129 | |
| BX950557.1 | - | 1 - 2 | 95 | 129 | ||
More Information
Set Membership: |
|

GATC
Genomic Map