Assay Details
| Gene Symbol: MSRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135558.2 | NP_001129030.1 | 2 - 3 | 237 | 65 | |
| NM_001346746.2 | NP_001333675.2 | 2 - 3 | 237 | 65 | ||
| GenBank mRNA | AJ454103.1 | - | 2 - 3 | 238 | 65 | |
| BG710256.1 | - | 2 - 3 | 217 | 65 | ||
| BM487648.1 | - | 2 - 3 | 218 | 65 | ||
| BU220886.1 | - | 2 - 3 | 205 | 65 | ||
| BX930278.2 | - | 2 - 3 | 180 | 65 | ||
| BX950711.1 | - | 2 - 3 | 229 | 65 | ||
| BX950828.2 | - | 2 - 3 | 205 | 65 | ||
| CD761648.1 | - | 2 - 3 | 254 | 65 | ||
More Information
Set Membership: |
|

MSRB1
Genomic Map