Assay Details
Gene Symbol: CZH5ORF34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015277557.2 | XP_015133043.1 | 6 - 7 | 3966 | 122 | |
XM_025145052.1 | XP_025000820.1 | 6 - 7 | 3966 | 122 | ||
XM_025145053.1 | XP_025000821.1 | 6 - 7 | 3966 | 122 | ||
XM_025145054.1 | XP_025000822.1 | 6 - 7 | 3966 | 122 | ||
XM_025145055.1 | XP_025000823.1 | 6 - 7 | 3966 | 122 | ||
XM_025145056.1 | XP_025000824.1 | 6 - 7 | 3966 | 122 | ||
GenBank mRNA | CR352435.1 | - | 1 - 2 | 62 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
