Assay Details
Gene Symbol: CSTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198648.2 | NP_001185577.1 | 1 - 2 | 146 | 112 | |
GenBank mRNA | BX932006.1 | - | 1 - 2 | 141 | 112 | |
BX934852.1 | - | 1 - 2 | 146 | 112 | ||
BX936150.1 | - | 1 - 2 | 144 | 112 | ||
CR352506.1 | - | 1 - 2 | 82 | 112 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001198648.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
