Assay Details
| Gene Symbol: EIF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025147517.1 | XP_025003285.1 | 2 - 3 | 272 | 82 | |
| GenBank mRNA | BX935434.1 | - | 2 - 3 | 305 | 82 | |
| CR352812.1 | - | 2 - 3 | 315 | 82 | ||
More Information
Set Membership: |
|

EIF1B
Genomic Map