Assay Details
| Gene Symbol: MRPS34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277597.1 | NP_001264526.1 | 1 - 2 | 344 | 154 | |
| GenBank mRNA | BX930865.2 | - | 1 - 2 | 353 | 154 | |
More Information
Set Membership: |
|

MRPS34
Genomic Map