Assay Details
Gene Symbol: WASHC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004937815.3 | XP_004937872.1 | 7 - 8 | 589 | 77 | |
XM_015287817.2 | XP_015143303.1 | 7 - 8 | 589 | 77 | ||
XM_015287824.2 | XP_015143310.1 | 7 - 8 | 589 | 77 | ||
XM_025155660.1 | XP_025011428.1 | 7 - 8 | 589 | 77 | ||
XM_025155663.1 | XP_025011431.1 | 7 - 8 | 583 | 77 | ||
XM_025155666.1 | XP_025011434.1 | 7 - 8 | 577 | 77 | ||
GenBank mRNA | BX931027.2 | - | 7 - 8 | 553 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
