Assay Details
| Gene Symbol: NSMCE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277463.1 | NP_001264392.1 | 6 - 7 | 668 | 85 | |
| NR_102407.1 | 5 - 6 | 561 | 85 | |||
| GenBank mRNA | BU104681.1 | - | 2 - 3 | 175 | 85 | |
| BX930919.2 | - | 6 - 7 | 671 | 85 | ||
| BX932130.1 | - | 5 - 6 | 561 | 85 | ||
| BX932910.2 | - | 6 - 7 | 661 | 85 | ||
More Information
Set Membership: |
|

NSMCE2
Genomic Map