Assay Details
Gene Symbol: SPSB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004947449.3 | XP_004947506.1 | 2 - 3 | 894 | 72 | |
XM_004947450.3 | XP_004947507.1 | 2 - 3 | 862 | 72 | ||
XM_025142572.1 | XP_024998340.1 | 2 - 3 | 1061 | 72 | ||
XM_417599.6 | XP_417599.1 | 2 - 3 | 860 | 72 | ||
GenBank mRNA | BX933393.2 | - | 1 - 2 | 839 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
