Assay Details
Gene Symbol: HDAC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277141.1 | NP_001264070.1 | 9 - 10 | 1012 | 71 | |
XM_015293066.2 | XP_015148552.1 | 10 - 11 | 1290 | 71 | ||
XM_025154462.1 | XP_025010230.1 | 10 - 11 | 1108 | 71 | ||
XM_025154463.1 | XP_025010231.1 | 9 - 10 | 844 | 71 | ||
GenBank mRNA | BX933925.2 | - | 5 - 6 | 443 | 71 | |
BX950678.1 | - | 8 - 9 | 874 | 71 | ||
CO635753.1 | - | 1 - 2 | 949 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
