Assay Details
| Gene Symbol: UBE2V1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205097.1 | NP_001192026.1 | 3 - 4 | 298 | 114 | |
| XM_015296313.2 | XP_015151799.2 | 3 - 4 | 333 | 114 | ||
| XM_015296314.2 | XP_015151800.2 | 3 - 4 | 350 | 114 | ||
| GenBank mRNA | BX933933.2 | - | 3 - 4 | 308 | 114 | |
| L77699.1 | - | 3 - 4 | 298 | 114 | ||
More Information
Set Membership: |
|

UBE2V1
Genomic Map