Assay Details
| Gene Symbol: LPAR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278049.1 | NP_001264978.1 | 2 | 1898 | 81 | |
| XM_015278219.2 | XP_015133705.1 | 2 | 1844 | 81 | ||
| XM_015278221.2 | XP_015133707.1 | 2 | 1772 | 81 | ||
| XM_025149545.1 | XP_025005313.1 | 3 | 2554 | 81 | ||
| GenBank mRNA | BX934031.2 | - | 1 | 1548 | 81 | |
| CR353562.1 | - | 2 | 1723 | 81 | ||
More Information
Set Membership: |
|

LPAR4
Genomic Map