Assay Details
| Gene Symbol: BCL11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003641409.4 | XP_003641457.1 | 2 - 3 | 1372 | 123 | |
| XM_004941933.3 | XP_004941990.1 | 2 - 3 | 1375 | 123 | ||
| GenBank mRNA | BX934432.2 | - | 2 - 3 | 449 | 123 | |
More Information
Set Membership: |
|

BCL11B
Genomic Map