Assay Details
Gene Symbol: GEMIN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BX929290.1 | - | 1 - 2 | 258 | 61 | |
BX930741.2 | - | 1 - 2 | 265 | 61 | ||
BX935317.2 | - | 1 - 2 | 287 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
