Assay Details
Gene Symbol: CMTM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199703.1 | NP_001186632.1 | 1 - 2 | 253 | 75 | |
XM_025147209.1 | XP_025002977.1 | 2 - 3 | 11172 | 75 | ||
GenBank mRNA | BU290096.1 | - | 1 - 2 | 164 | 75 | |
BX935400.2 | - | 1 - 2 | 253 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
