Assay Details
Gene Symbol: CMTM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199703.1 | NP_001186632.1 | 2 - 3 | 425 | 71 | |
XM_025147209.1 | XP_025002977.1 | 3 - 4 | 11344 | 71 | ||
GenBank mRNA | BU290096.1 | - | 2 - 3 | 336 | 71 | |
BX935400.2 | - | 2 - 3 | 425 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
