Assay Details
| Gene Symbol: POLR2E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_418224.6 | XP_418224.2 | 3 - 4 | 378 | 67 | |
| GenBank mRNA | BX929663.1 | - | 3 - 4 | 362 | 67 | |
| BX930905.2 | - | 3 - 4 | 361 | 67 | ||
| BX935423.2 | - | 3 - 4 | 365 | 67 | ||
| BX935442.2 | - | 3 - 4 | 353 | 67 | ||
| CR385484.1 | - | 3 - 4 | 341 | 67 | ||
More Information
Set Membership: |
|

POLR2E
Genomic Map