Assay Details
Gene Symbol: RAB40B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278050.3 | NP_001264979.1 | 5 - 6 | 911 | 55 | |
GenBank mRNA | BX950715.2 | - | 5 - 6 | 911 | 55 | |
CR406234.1 | - | 3 - 4 | 364 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
