Assay Details
| Gene Symbol: AEBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_416415.6 | XP_416415.5 | 1 - 2 | 753 | 72 | |
| GenBank mRNA | CR353321.1 | - | 1 - 2 | 68 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AEBP2| Gene Symbol: AEBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_416415.6 | XP_416415.5 | 1 - 2 | 753 | 72 | |
| GenBank mRNA | CR353321.1 | - | 1 - 2 | 68 | 72 | |
Set Membership: |
|