Assay Details
| Gene Symbol: ACYP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167762.1 | NP_001161234.1 | 2 - 3 | 164 | 66 | |
| XM_025148627.1 | XP_025004395.1 | 2 - 3 | 220 | 66 | ||
| GenBank mRNA | BG710739.1 | - | 2 - 3 | 139 | 66 | |
| BU393960.1 | - | 2 - 3 | 164 | 66 | ||
| CR353528.1 | - | 2 - 3 | 162 | 66 | ||
More Information
Set Membership: |
|

ACYP2
Genomic Map