Assay Details
Gene Symbol: COQ7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277172.1 | NP_001264101.1 | 4 - 5 | 546 | 88 | |
XM_015294222.2 | XP_015149708.1 | 4 - 5 | 610 | 88 | ||
XM_015294223.2 | XP_015149709.1 | 4 - 5 | 610 | 88 | ||
XM_025155105.1 | XP_025010873.1 | 4 - 5 | 610 | 88 | ||
XM_025155106.1 | XP_025010874.1 | 4 - 5 | 610 | 88 | ||
GenBank mRNA | BX933254.1 | - | 4 - 5 | 544 | 88 | |
BX935091.1 | - | 4 - 5 | 541 | 88 | ||
BX935416.1 | - | 4 - 5 | 551 | 88 | ||
CR385437.1 | - | 4 - 5 | 546 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
