Assay Details
Gene Symbol: RSRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025153635.1 | XP_025009403.1 | 2 - 3 | 308 | 86 | |
XM_025153636.1 | XP_025009404.1 | 2 - 3 | 290 | 86 | ||
GenBank mRNA | BX934438.2 | - | 2 - 3 | 260 | 86 | |
CR385561.1 | - | 2 - 3 | 251 | 86 | ||
CR407368.1 | - | 2 - 3 | 275 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
