Assay Details
Gene Symbol: RALYL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199478.1 | NP_001186407.1 | 2 - 3 | 177 | 75 | |
XM_015282796.2 | XP_015138282.1 | 4 - 5 | 537 | 75 | ||
XM_015282797.2 | XP_015138283.1 | 5 - 6 | 644 | 75 | ||
XM_015282798.2 | XP_015138284.1 | 4 - 5 | 528 | 75 | ||
XM_015282800.2 | XP_015138286.1 | 3 - 4 | 1998 | 75 | ||
XM_015282801.2 | XP_015138287.1 | 4 - 5 | 537 | 75 | ||
XM_015282802.2 | XP_015138288.1 | 4 - 5 | 537 | 75 | ||
XM_015282803.2 | XP_015138289.1 | 4 - 5 | 537 | 75 | ||
XM_015282804.2 | XP_015138290.1 | 2 - 3 | 214 | 75 | ||
XM_015282805.2 | XP_015138291.1 | 3 - 4 | 652 | 75 | ||
XM_015282806.2 | XP_015138292.1 | 3 - 4 | 634 | 75 | ||
XM_025147092.1 | XP_025002860.1 | 4 - 5 | 631 | 75 | ||
XM_025147093.1 | XP_025002861.1 | 3 - 4 | 419 | 75 | ||
XM_025147094.1 | XP_025002862.1 | 4 - 5 | 537 | 75 | ||
XM_025147095.1 | XP_025002863.1 | 2 - 3 | 204 | 75 | ||
GenBank mRNA | CR385662.1 | - | 2 - 3 | 177 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
