Assay Details
| Gene Symbol: GLUD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015288033.2 | XP_015143519.1 | 3 - 4 | 602 | 78 | |
| GenBank mRNA | CR387697.1 | - | 3 - 4 | 450 | 78 | |
| MF415491.1 | - | 3 - 4 | 377 | 78 | ||
More Information
Set Membership: |
|

GLUD2
Genomic Map