Assay Details
| Gene Symbol: XIRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204348.3 | NP_989679.3 | 3 - 4 | 371 | 100 | |
| XM_015281203.2 | XP_015136689.1 | 7 - 8 | 1015 | 100 | ||
| GenBank mRNA | AF051944.2 | - | 3 - 4 | 337 | 100 | |
| BX932230.1 | - | 3 - 4 | 371 | 100 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_204348.3 transcripts.
Set Membership: |
|

XIRP1
Genomic Map