Assay Details
| Gene Symbol: MRPS11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005821.1 | NP_001005821.1 | 4 - 5 | 415 | 56 | |
| GenBank mRNA | AJ719914.1 | - | 4 - 5 | 415 | 56 | |
More Information
Set Membership: |
|

MRPS11
Genomic Map