Assay Details
| Gene Symbol: KCTD14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277837.1 | NP_001264766.1 | 1 - 2 | 112 | ||
| GenBank mRNA | BU450487.1 | - | 1 - 2 | 112 | ||
| BX929662.2 | - | 1 - 2 | 164 | 112 | ||
| BX930218.2 | - | 1 - 2 | 202 | 112 | ||
| BX932683.2 | - | 1 - 2 | 138 | 112 | ||
| BX950733.1 | - | 1 - 2 | 112 | |||
More Information
Set Membership: |
|

KCTD14
Genomic Map