Assay Details
| Gene Symbol: YAE1D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_418862.6 | XP_418862.1 | 2 - 3 | 372 | 124 | |
| GenBank mRNA | BX931906.2 | - | 2 - 3 | 347 | 124 | |
| BX935780.2 | - | 1 - 2 | 244 | 124 | ||
| Gene Symbol: RALA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025147578.1 | XP_025003346.1 | 2 - 3 | 369 | 124 | |
| XM_025147579.1 | XP_025003347.1 | 2 - 3 | 369 | 124 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025147578.1, XM_025147579.1 transcripts.
Set Membership: |
|

YAE1D1
Genomic Map