Assay Details
| Gene Symbol: YAE1D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_418862.6 | XP_418862.1 | 3 - 4 | 496 | 117 | |
| GenBank mRNA | BX931906.2 | - | 3 - 4 | 471 | 117 | |
More Information
Set Membership: |
|

YAE1D1
Genomic Map