Assay Details
Gene Symbol: RRAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277606.2 | NP_001264535.2 | 4 - 5 | 677 | 65 | |
GenBank mRNA | BX934073.1 | - | 4 - 5 | 678 | 65 | |
BX950258.2 | - | 4 - 5 | 679 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
