Assay Details
| Gene Symbol: DYDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199517.1 | NP_001186446.1 | 3 - 4 | 352 | 139 | |
| GenBank mRNA | BX950529.2 | - | 3 - 4 | 340 | 139 | |
More Information
Set Membership: |
|

DYDC2
Genomic Map