Assay Details
Gene Symbol: LOC424577 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_422416.6 | XP_422416.3 | 3 | 1494 | 159 | |
GenBank mRNA | CR388642.1 | - | 1 | 144 | 159 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
