Assay Details
| Gene Symbol: RPS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252126.1 | NP_001239055.1 | 4 - 5 | 90 | ||
| GenBank mRNA | AY389966.1 | - | 3 - 4 | 90 | ||
| CF250737.1 | - | 4 - 5 | 435 | 90 | ||
| CN218040.1 | - | 3 - 4 | 90 | |||
| CR390133.1 | - | 3 - 4 | 392 | 90 | ||
| CR390136.1 | - | 3 - 4 | 90 | |||
| CR390875.1 | - | 4 - 5 | 90 | |||
More Information
Set Membership: |
|

RPS8
Genomic Map