Assay Details
| Gene Symbol: SLC16A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321736.1 | NP_001308665.1 | 2 - 3 | 84 | ||
| XM_025148740.1 | XP_025004508.1 | 2 - 3 | 321 | 84 | ||
| GenBank mRNA | CR390252.1 | - | 2 - 3 | 305 | 84 | |
| KT876068.1 | - | 2 - 3 | 84 | |||
More Information
Set Membership: |
|

SLC16A10
Genomic Map